Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UTP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016037.3 | NP_057121.2 | 3 | 270 | ||
XM_011541555.2 | XP_011539857.1 | 2 | 204 | |||
GenBank mRNA | AF067802.1 | - | 3 | 233 | ||
AF151852.1 | - | 3 | 248 | |||
AK023587.1 | - | 1 | 1139 | |||
AK291900.1 | - | 3 | 200 | |||
AK296018.1 | - | 3 | 203 | |||
AK308377.1 | - | 2 | 205 | |||
BC005182.1 | - | 3 | 215 | 3995984 | ||
BU940926.1 | - | 3 | 273 |