Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXNDC11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303447.1 | NP_001290376.1 | 2 | 587 | ||
NM_001324022.1 | NP_001310951.1 | 2 | 587 | |||
NM_001324024.1 | NP_001310953.1 | 2 | 587 | |||
NM_001324025.1 | NP_001310954.1 | 2 | 587 | |||
NM_015914.6 | NP_056998.4 | 2 | 587 | |||
NR_136671.1 | 2 | 587 | ||||
NR_136672.1 | 2 | 587 | ||||
NR_136673.1 | 2 | 587 | ||||
NR_136674.1 | 2 | 587 | ||||
XM_011522515.2 | XP_011520817.1 | 2 | 544 | |||
XM_011522516.2 | XP_011520818.1 | 2 | 434 | |||
XM_017023268.1 | XP_016878757.1 | 2 | 544 | |||
GenBank mRNA | AK027646.1 | - | 2 | 501 | ||
BC002856.1 | - | 2 | 556 | 3635977 | ||
BC018635.2 | - | 2 | 557 | |||
BX648407.1 | - | 3 | 294 | |||
CN347909.1 | - | 2 | 587 |