Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CUTA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014433.2 | NP_001014433.1 | 4, 5 | 522 | ||
NM_001014837.1 | NP_001014837.1 | 4, 5 | 771 | |||
NM_001014838.1 | NP_001014838.1 | 4, 5 | 740 | |||
NM_001014840.1 | NP_001014840.1 | 4, 5 | 650 | |||
NM_015921.2 | NP_057005.1 | 3, 4 | 913 | |||
GenBank mRNA | AF106943.1 | - | 3, 4 | 550 | ||
AF230924.1 | - | 3, 4 | 336 | |||
BC005890.2 | - | 4, 5 | 347 | 3505574 | ||
BC107751.1 | - | 3, 4 | 550 | 6714433 | ||
CN363642.1 | - | 4, 5 | 552 |