Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPH5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077394.1 | NP_001070862.1 | 8 | 1454 | ||
NM_001077395.1 | NP_001070863.1 | 8 | 1433 | |||
NM_015958.2 | NP_057042.2 | 8 | 1436 | |||
XM_005270938.1 | XP_005270995.1 | 7 | 1332 | |||
XM_005270939.4 | XP_005270996.1 | 7 | 1244 | |||
XM_005270941.3 | XP_005270998.1 | 7 | 1219 | |||
XM_006710683.3 | XP_006710746.1 | 6 | 1091 | |||
XM_011541569.1 | XP_011539871.1 | 7 | 1329 | |||
XM_011541570.2 | XP_011539872.1 | 7 | 1236 | |||
XM_017001460.1 | XP_016856949.1 | 8 | 1456 | |||
XM_017001461.1 | XP_016856950.1 | 7 | 1243 | |||
XM_017001462.1 | XP_016856951.1 | 7 | 1238 | |||
GenBank mRNA | AF132964.1 | - | 8 | 1441 | ||
AK125485.1 | - | 9 | 1805 | |||
AK289351.1 | - | 8 | 1422 | |||
BM808505.1 | - | 1 | 253 |