Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIDT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040455.1 | NP_001035545.1 | 5 | 590 | ||
GenBank mRNA | AF151799.1 | - | 5 | 632 | ||
AK292501.1 | - | 5 | 1070 | |||
AK292974.1 | - | 5 | 581 | |||
AK303023.1 | - | 5 | 546 | |||
AY358442.1 | - | 5 | 599 | |||
BC114522.1 | - | 5 | 619 | 40035836 | ||
KC109742.1 | - | 5 | 546 |