Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001032391.1 | NP_001027563.1 | 3 | 412 | ||
NM_016309.2 | NP_057393.2 | 3 | 412 | |||
XM_005255354.3 | XP_005255411.1 | 3 | 201 | |||
XM_011545862.2 | XP_011544164.1 | 4 | 526 | |||
XM_011545864.1 | XP_011544166.1 | 4 | 526 | |||
GenBank mRNA | AF037601.1 | - | 3 | 288 | ||
AF151826.1 | - | 3 | 396 | |||
AK025884.1 | - | 4 | 519 | |||
AK223364.1 | - | 4 | 570 | |||
AK225271.1 | - | 3 | 356 | |||
AK291885.1 | - | 3 | 356 | |||
AK314409.1 | - | 3 | 357 | |||
AL137283.1 | - | 3 | 216 | |||
AL560574.3 | - | 3 | 403 | |||
BC001214.1 | - | 3 | 379 | 3533656 | ||
BC014217.2 | - | 4 | 500 | 3959936 | ||
BI459868.1 | - | 3 | 434 |