Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBMX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016024.2 | NP_057108.2 | 4 | 304 | ||
GenBank mRNA | AF078865.1 | - | 3 | 240 | ||
AF151837.1 | - | 3 | 240 | |||
AK292855.1 | - | 4 | 286 | |||
AL050405.1 | - | 4 | 279 | |||
BC003676.1 | - | 4 | 304 | |||
BC033750.1 | - | 4 | 272 | 5210341 | ||
BC040433.1 | - | 4 | 255 | |||
BC105582.1 | - | 4 | 291 |