Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDHHC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008222.2 | NP_001008223.1 | 2 | 525 | ||
NM_016032.3 | NP_057116.2 | 3 | 505 | |||
XM_011531348.2 | XP_011529650.1 | 3 | 492 | |||
GenBank mRNA | AB209807.1 | - | 2 | 444 | ||
AF151847.1 | - | 3 | 393 | |||
AK001112.1 | - | 3 | 455 | |||
AK001524.1 | - | 2 | 256 | |||
AL161962.1 | - | 3 | 382 | |||
AY358558.1 | - | 3 | 437 | |||
AY952881.8 | - | 3 | 477 | |||
BC000035.2 | - | 2 | 399 | 3506601 | ||
BC003128.1 | - | 3 | 443 | 2957911 | ||
BC006200.2 | - | 3 | 439 | 2964425 | ||
BC012826.2 | - | 3 | 439 | 2964425 | ||
BM909699.1 | - | 432 | ||||
CD627424.1 | - | 3 | 470 | |||
DA493628.1 | - | 3 | 505 | |||
DA768650.1 | - | 2 | 525 |