Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127713.1 | NP_001121185.1 | 5, 6 | 955 | ||
NM_015915.4 | NP_056999.2 | 4, 5 | 771 | |||
NM_181598.3 | NP_853629.2 | 4, 5 | 771 | |||
GenBank mRNA | AF131801.1 | - | 3, 4 | 295 | ||
AF444143.1 | - | 4, 5 | 702 | |||
AK223436.1 | - | 4, 5 | 684 | |||
AK290185.1 | - | 4, 5 | 684 | |||
AK312518.1 | - | 4, 5 | 660 | |||
AL833591.1 | - | 5, 6 | 739 | |||
AY032844.1 | - | 4, 5 | 699 | |||
BC010708.2 | - | 4, 5 | 674 | 3869877 | ||
CR457153.1 | - | 3, 4 | 281 | |||
KU178621.1 | - | 4, 5 | 530 | |||
KU178622.1 | - | 4, 5 | 530 | |||
KU178623.1 | - | 4, 5 | 530 |