Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF077202.1 | - | 3 | 127 | ||
AF161448.1 | - | 2 | 76 | |||
AK126594.1 | - | 3 | 89 | |||
AL712164.1 | - | 3 | 104 | |||
AY508979.1 | - | 1 | 110 | |||
AY794023.1 | - | 3 | 111 | |||
BC001102.2 | - | 1 | 99 | |||
BC080531.1 | - | 3 | 112 | 4875991 | ||
BU536461.1 | - | 3 | 156 |