Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METTL13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007239.1 | NP_001007240.1 | 7 | 1687 | ||
NM_014955.2 | NP_055770.1 | 7 | 1830 | |||
NM_015935.4 | NP_057019.3 | 7 | 2155 | |||
XM_017001455.1 | XP_016856944.1 | 6 | 1649 | |||
XM_017001456.1 | XP_016856945.1 | 6 | 1649 | |||
XM_017001457.1 | XP_016856946.1 | 5 | 1430 | |||
XM_017001458.1 | XP_016856947.1 | 6 | 2107 | |||
XM_017001459.1 | XP_016856948.1 | 6 | 1657 | |||
GenBank mRNA | AB020666.2 | - | 7 | 1910 | ||
AB242174.1 | - | 7 | 2080 | |||
AB242175.1 | - | 7 | 2068 | |||
AF132936.1 | - | 7 | 2080 | |||
AK001172.1 | - | 5 | 842 | |||
AK027552.1 | - | 7 | 2074 | |||
AK027621.1 | - | 7 | 1769 | |||
AK027886.1 | - | 6 | 1626 | |||
AK074552.1 | - | 3 | 908 | |||
AK129561.1 | - | 6 | 1503 | |||
AK223493.1 | - | 7 | 2116 | |||
AK291740.1 | - | 7 | 2102 | |||
AK304471.1 | - | 8 | 1936 | |||
AL049669.1 | - | 7 | 1612 | |||
BC006814.1 | - | 4 | 705 | |||
BC029083.1 | - | 7 | 1909 | 4509086 |