Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NMD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015938.4 | NP_057022.2 | 16 | 2182 | ||
XM_005247511.1 | XP_005247568.1 | 16 | 2132 | |||
XM_005247512.1 | XP_005247569.1 | 16 | 2463 | |||
GenBank mRNA | AK024706.1 | - | 3 | 781 | ||
AK094749.1 | - | 15 | 2485 | |||
BC013317.1 | - | 16 | 2165 | 4291797 |