Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEMO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001137602.2 | NP_001131074.1 | 6 | 943 | ||
NM_001301833.1 | NP_001288762.1 | 8 | 723 | |||
NM_001301852.1 | NP_001288781.1 | 6 | 502 | |||
NM_015955.3 | NP_057039.1 | 7 | 1012 | |||
XM_011532892.2 | XP_011531194.1 | 8 | 660 | |||
XM_011532896.2 | XP_011531198.1 | 5 | 395 | |||
XM_017004255.1 | XP_016859744.1 | 8 | 633 | |||
GenBank mRNA | AB041018.1 | - | 7 | 1012 | ||
AB041019.1 | - | 7 | 661 | |||
AF132961.1 | - | 7 | 710 | |||
AF363446.1 | - | 7 | 929 | |||
AF529368.1 | - | 7 | 595 | |||
AK290753.1 | - | 8 | 679 | |||
AK297128.1 | - | 6 | 654 | |||
BC018733.1 | - | 7 | 977 | 4851517 | ||
BC070046.1 | - | 8 | 752 | 5273070 | ||
BC094681.1 | - | 7 | 616 | 6205399 | ||
BG033287.1 | - | 6 | 451 |