Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THAP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164356.1 | NP_001157828.1 | 5 | 703 | ||
NM_015963.5 | NP_057047.4 | 6 | 2146 | |||
XM_005247016.4 | XP_005247073.3 | 6 | 2460 | |||
XM_011511291.2 | XP_011509593.1 | 6 | 2029 | |||
XM_017004256.1 | XP_016859745.1 | 6 | 1940 | |||
GenBank mRNA | AF132970.1 | - | 5 | 702 | ||
AF258556.1 | - | 6 | 1898 | |||
AK001216.1 | - | 5 | 1193 | |||
AK225608.1 | - | 5 | 671 | |||
BC000247.1 | - | 5 | 1588 | |||
BC000767.2 | - | 4 | 910 | |||
BC001842.2 | - | 4 | 910 | |||
BC009439.1 | - | 5 | 1684 | |||
BC069235.1 | - | 6 | 2076 | 6645198 | ||
BC071896.1 | - | 5 | 1112 | |||
BC094822.1 | - | 5 | 1112 |