Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRELID3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256403.1 | NP_001243332.1 | 5 | 1261 | ||
NM_016045.2 | NP_057129.2 | 6 | 1351 | |||
GenBank mRNA | AF151865.1 | - | 6 | 1321 | ||
BC010649.1 | - | 6 | 1301 | 3873112 | ||
BC013969.2 | - | 6 | 1313 | 3631601 |