Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016050.4 | NP_057134.1 | 2 | 255 | ||
NM_170738.3 | NP_733934.1 | 2 | 177 | |||
NM_170739.3 | NP_733935.1 | 2 | 255 | |||
NR_120588.1 | 2 | 255 | ||||
NR_120589.1 | 2 | 105 | ||||
XM_017018135.1 | XP_016873624.1 | 2 | 189 | |||
GenBank mRNA | AB049638.1 | - | 2 | 227 | ||
AF151871.1 | - | 2 | 239 | |||
AK127603.1 | - | 2 | 864 | |||
AK223112.1 | - | 2 | 231 | |||
AK290084.1 | - | 2 | 233 | |||
BC005002.1 | - | 2 | 234 | 3542758 | ||
BC108277.1 | - | 2 | 150 | 4416083 | ||
BI193255.1 | - | 2 | 169 | |||
BM554886.1 | - | 2 | 217 | |||
BM755421.1 | - | 2 | 246 | |||
BU597133.1 | - | 2 | 258 |