Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM96B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016062.3 | NP_057146.1 | 2 | 256 | ||
NR_024525.2 | 1 | 412 | ||||
NR_046109.1 | 2 | 322 | ||||
GenBank mRNA | AF151886.1 | - | 2 | 232 | ||
AF161467.1 | - | 2 | 221 | |||
BC001733.1 | - | 2 | 216 | 3353705 | ||
BC005023.1 | - | 2 | 219 | 3632169 | ||
BI598300.1 | - | 1 | 407 | |||
BI603322.1 | - | 2 | 286 | |||
BM091095.1 | - | 1 | 336 | |||
BQ955804.1 | - | 2 | 188 | |||
CD674578.1 | - | 2 | 240 | |||
DA270099.1 | - | 1 | 413 | |||
DR155511.1 | - | 1 | 453 |