Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS18C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297767.1 | NP_001284696.1 | 2 | 269 | ||
NM_001297769.1 | NP_001284698.1 | 2 | 269 | |||
NM_001297770.1 | NP_001284699.1 | 2 | 269 | |||
NM_016067.3 | NP_057151.1 | 2 | 269 | |||
GenBank mRNA | AF151892.1 | - | 2 | 182 | ||
AK002008.1 | - | 2 | 135 | |||
AK026620.1 | - | 2 | 236 | |||
BC005186.1 | - | 2 | 160 | 3839381 | ||
BE874586.1 | - | 2 | 111 | |||
CA455563.1 | - | 1 | 146 | |||
CN262115.1 | - | 2 | 183 | |||
DA446717.1 | - | 2 | 270 |