Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270879.1 | NP_001257808.1 | 6 | 702 | ||
NM_016098.3 | NP_057182.1 | 5 | 486 | |||
NR_073093.1 | 6 | 715 | ||||
XM_011535894.1 | XP_011534196.1 | 5 | 1485 | |||
XM_011535895.1 | XP_011534197.1 | 4 | 1269 | |||
GenBank mRNA | AF125101.1 | - | 5 | 472 | ||
AF151887.1 | - | 5 | 468 | |||
AF275811.1 | - | 3 | 193 | |||
AL556427.3 | - | 5 | 405 | |||
AL561589.3 | - | 6 | 590 | |||
BC000810.1 | - | 5 | 396 | 3452973 | ||
BG828174.1 | - | 5 | 444 |