Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOLGA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002296.1 | NP_001002296.1 | 2 | 411 | ||
NM_001174124.1 | NP_001167595.1 | 3 | 288 | |||
NM_016099.2 | NP_057183.2 | 3 | 288 | |||
GenBank mRNA | AB104615.1 | - | 3 | 268 | ||
AF068291.1 | - | 2 | 242 | |||
AF125102.1 | - | 3 | 252 | |||
AK026613.1 | - | 3 | 268 | |||
AL695304.1 | - | 3 | 297 | |||
BC012032.1 | - | 2 | 411 | 4707385 | ||
BI553491.1 | - | 3 | 290 |