Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199434.1 | NP_001186363.1 | 4 | 745 | ||
NM_016101.4 | NP_057185.1 | 5 | 886 | |||
GenBank mRNA | AB062398.1 | - | 5 | 588 | ||
AB062479.1 | - | 5 | 588 | |||
AF085360.1 | - | 5 | 601 | |||
AF132971.1 | - | 5 | 601 | |||
AF161528.1 | - | 4 | 459 | |||
AK001158.1 | - | 4 | 437 | |||
BC015941.1 | - | 5 | 613 | 3922688 |