Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SARAF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016127.5 | NP_057211.4 | 2 | 472 | ||
XM_017013567.1 | XP_016869056.1 | 2 | 722 | |||
XM_017013568.1 | XP_016869057.1 | 2 | 150 | |||
XM_017013569.1 | XP_016869058.1 | 2 | 150 | |||
GenBank mRNA | AB028926.1 | - | 2 | 332 | ||
AF078855.1 | - | 2 | 300 | |||
AF100748.1 | - | 2 | 229 | |||
AF490252.1 | - | 2 | 213 | |||
AK025927.1 | - | 2 | 382 | |||
AK075340.1 | - | 2 | 330 | |||
AK130057.1 | - | 1 | 80 | |||
AK222632.1 | - | 2 | 229 | |||
AK295360.1 | - | 3 | 277 | |||
AK314659.1 | - | 2 | 355 | |||
AY359104.1 | - | 2 | 397 | |||
BC015012.1 | - | 2 | 301 | 3908758 | ||
DC359554.1 | - | 2 | 473 | |||
JQ348891.1 | - | 2 | 341 | |||
Z70223.1 | - | 2 | 239 |