Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COPS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258006.1 | NP_001244935.1 | 3, 4 | 447 | ||
NM_016129.2 | NP_057213.2 | 3, 4 | 447 | |||
XM_011532019.1 | XP_011530321.1 | 3, 4 | 443 | |||
GenBank mRNA | AF100757.1 | - | 3, 4 | 311 | ||
AK001148.1 | - | 3, 4 | 443 | |||
AK001210.1 | - | 3, 4 | 274 | |||
AK024005.1 | - | 3, 4 | 332 | |||
AK094238.1 | - | 3, 4 | 1085 | |||
AK223178.1 | - | 3, 4 | 317 | |||
BC004302.1 | - | 3, 4 | 310 | 3623514 | ||
BC009292.2 | - | 3, 4 | 310 | 4111685 | ||
BC093007.1 | - | 3, 4 | 332 | 30340740 | ||
CD690475.1 | - | 3, 4 | 478 | |||
KU178633.1 | - | 3, 4 | 308 | |||
KU178634.1 | - | 3, 4 | 308 |