Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSTA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000847.4 | NP_000838.3 | 7 | 772 | ||
XM_006715072.3 | XP_006715135.1 | 5 | 606 | |||
XM_011514533.2 | XP_011512835.1 | 4 | 598 | |||
XM_017010794.1 | XP_016866283.1 | 5 | 647 | |||
GenBank mRNA | AF020919.1 | - | 6 | 632 | ||
BC020619.1 | - | 7 | 759 | 4721622 |