Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COQ7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190983.1 | NP_001177912.1 | 2 | 139 | ||
NM_016138.4 | NP_057222.2 | 2 | 190 | |||
GenBank mRNA | AF098948.1 | - | 2 | 148 | ||
AK024291.1 | - | 2 | 171 | |||
AK225115.1 | - | 2 | 139 | |||
AK315470.1 | - | 2 | 190 | |||
AL136647.1 | - | 2 | 142 | |||
BC003185.1 | - | 2 | 181 | 3503763 | ||
BC043272.1 | - | 2 | 940 | 5297074 |