Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR83OS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016145.3 | NP_057229.1 | 3 | 481 | ||
XM_017026864.1 | XP_016882353.1 | 4 | 1475 | |||
GenBank mRNA | AF059620.1 | - | 3 | 249 | ||
AF078861.1 | - | 3 | 466 | |||
AF151898.1 | - | 3 | 465 | |||
AK091030.1 | - | 3 | 2186 | |||
AK311944.1 | - | 3 | 245 | |||
BC001192.1 | - | 3 | 229 | |||
BC042919.1 | - | 3 | 250 | 5932109 | ||
BC061912.1 | - | 3 | 250 | 4444565 |