Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSTM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000849.4 | NP_000840.2 | 8 | 876 | ||
NR_024537.1 | 7 | 800 | ||||
GenBank mRNA | AB209816.1 | - | 7 | 585 | ||
AK304300.1 | - | 6 | 528 | |||
AK313444.1 | - | 7 | 594 | |||
BC000088.2 | - | 7 | 581 | 3509784 | ||
BC008790.2 | - | 8 | 876 | 3630923 | ||
BT019945.1 | - | 7 | 566 | |||
CR542108.1 | - | 7 | 566 | |||
J05459.1 | - | 7 | 583 |