Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRTO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016183.3 | NP_057267.2 | 3 | 409 | ||
XM_006710675.3 | XP_006710738.1 | 3 | 567 | |||
GenBank mRNA | AF173378.1 | - | 3 | 184 | ||
AK024227.1 | - | 3 | 170 | |||
AK027569.1 | - | 3 | 185 | |||
AK295328.1 | - | 3 | 156 | |||
AY303790.1 | - | 3 | 165 | |||
BC003013.1 | - | 3 | 183 | 2821148 | ||
BC006504.2 | - | 3 | 183 | 2821148 | ||
BP312953.1 | - | 3 | 409 | |||
BQ670859.1 | - | 3 | 364 |