Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100607.2 | NP_001094077.1 | 2 | 318 | ||
NM_016186.2 | NP_057270.1 | 2 | 651 | |||
XM_005267733.4 | XP_005267790.1 | 2 | 403 | |||
XM_017021353.1 | XP_016876842.1 | 3 | 602 | |||
GenBank mRNA | AF181467.1 | - | 2 | 651 | ||
BC022261.1 | - | 2 | 318 | 4710734 | ||
BX248011.1 | - | 2 | 305 |