Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPRKB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016058.2 | NP_057142.1 | 4 | 392 | ||
XM_005264343.3 | XP_005264400.1 | 5 | 1168 | |||
XM_006712026.3 | XP_006712089.1 | 6 | 983 | |||
XM_006712027.3 | XP_006712090.1 | 5 | 1097 | |||
XM_011532877.2 | XP_011531179.1 | 5 | 1094 | |||
XM_011532878.2 | XP_011531180.1 | 5 | 441 | |||
XM_011532879.2 | XP_011531181.1 | 5 | 755 | |||
XM_011532880.2 | XP_011531182.1 | 5 | 752 | |||
XM_017004232.1 | XP_016859721.1 | 6 | 981 | |||
XM_017004233.1 | XP_016859722.1 | 5 | 1178 | |||
XM_017004234.1 | XP_016859723.1 | 4 | 1064 | |||
XM_017004235.1 | XP_016859724.1 | 5 | 846 | |||
GenBank mRNA | AF060921.1 | - | 4 | 331 | ||
AF151879.1 | - | 4 | 387 | |||
AK097254.1 | - | 3 | 1944 | |||
AK291071.1 | - | 4 | 323 | |||
AK295239.1 | - | 4 | 269 | |||
AY157986.1 | - | 4 | 398 | |||
AY157987.1 | - | 3 | 182 | |||
AY643713.1 | - | 4 | 343 | |||
BC029492.1 | - | 4 | 344 | 5271898 | ||
DA688967.1 | - | 4 | 392 |