Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016065.3 | NP_057149.1 | 3 | 554 | ||
GenBank mRNA | AB049948.1 | - | 3 | 440 | ||
AF151890.1 | - | 3 | 507 | |||
AK025715.1 | - | 3 | 487 | |||
AK098291.1 | - | 3 | 500 | |||
BC021106.1 | - | 3 | 449 | 5020046 |