Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDGFRP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016073.3 | NP_057157.1 | 3 | 850 | ||
XM_006720554.3 | XP_006720617.1 | 3 | 386 | |||
GenBank mRNA | AB029156.1 | - | 3 | 366 | ||
AF110642.1 | - | 3 | 370 | |||
AF151900.1 | - | 3 | 370 | |||
AK001280.1 | - | 3 | 401 | |||
BC015483.1 | - | 3 | 344 | 3913558 | ||
BC040554.1 | - | 3 | 853 |