Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COMMD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016094.3 | NP_057178.2 | 5 | 485 | ||
NR_135149.1 | 5 | 565 | ||||
GenBank mRNA | AF059618.1 | - | 5 | 461 | ||
AF125096.1 | - | 5 | 472 | |||
AY542158.1 | - | 5 | 430 | |||
BC001228.1 | - | 5 | 464 | |||
BC022206.1 | - | 5 | 469 | |||
BC046131.1 | - | 5 | 447 | 4443942 | ||
BC093077.1 | - | 5 | 475 | 30372667 | ||
BI792102.1 | - | 5 | 493 | |||
BQ216242.1 | - | 2 | 228 | |||
CN483677.1 | - | 5 | 516 | |||
DA811158.1 | - | 5 | 484 |