Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF706 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042510.1 | NP_001035975.1 | 4 | 648 | ||
NM_001267708.1 | NP_001254637.1 | 5 | 692 | |||
NM_001267709.1 | NP_001254638.1 | 3 | 446 | |||
NM_016096.3 | NP_057180.1 | 3 | 533 | |||
GenBank mRNA | AF125099.1 | - | 3 | 369 | ||
AF275802.1 | - | 3 | 414 | |||
AF275809.1 | - | 3 | 414 | |||
AK027802.1 | - | 3 | 1757 | |||
AK290477.1 | - | 3 | 417 | |||
BC015925.1 | - | 1 | 290 | 3909510 | ||
BC068524.1 | - | 3 | 417 | 5263977 | ||
BG704587.1 | - | 3 | 451 | |||
BI755311.1 | - | 4 | 556 |