Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042399.1 | NP_001035858.1 | 6 | 1105 | ||
NM_016122.2 | NP_057206.2 | 7 | 1158 | |||
XM_006719437.2 | XP_006719500.1 | 6 | 1071 | |||
XM_011538424.1 | XP_011536726.1 | 8 | 1204 | |||
XM_011538427.2 | XP_011536729.1 | 7 | 930 | |||
XM_011538429.2 | XP_011536731.1 | 3 | 401 | |||
XM_017019385.1 | XP_016874874.1 | 7 | 1152 | |||
XM_017019386.1 | XP_016874875.1 | 7 | 1152 | |||
XM_017019387.1 | XP_016874876.1 | 6 | 909 | |||
XM_017019388.1 | XP_016874877.1 | 6 | 2774 | |||
XM_017019389.1 | XP_016874878.1 | 8 | 984 | |||
GenBank mRNA | AF155115.1 | - | 7 | 874 | ||
AK056316.1 | - | 6 | 1001 | |||
BC107746.1 | - | 4 | 419 | |||
BC125086.1 | - | 6 | 795 | |||
BC125087.1 | - | 7 | 708 | 40117807 |