Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAPPC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318488.1 | NP_001305417.1 | 3 | 732 | ||
NM_001318490.1 | NP_001305419.1 | 3 | 603 | |||
NM_001318492.1 | NP_001305421.1 | 3 | 839 | |||
NM_016146.5 | NP_057230.1 | 3 | 732 | |||
GenBank mRNA | AF078862.1 | - | 3 | 662 | ||
AF105025.1 | - | 3 | 670 | |||
AF151862.1 | - | 3 | 668 | |||
AF161520.1 | - | 3 | 455 | |||
AK025623.1 | - | 3 | 435 | |||
AK290520.1 | - | 3 | 435 | |||
AK293914.1 | - | 3 | 435 | |||
AK293977.1 | - | 3 | 435 | |||
BC010866.1 | - | 3 | 431 | 3876778 | ||
BF971819.1 | - | 3 | 518 | |||
BP332308.1 | - | 3 | 306 |