Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB4B-EGLN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037791.1 | 3 | 270 | |||
GenBank mRNA | AK291385.1 | - | 3 | 218 |
Gene Symbol: MIA-RAB4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037775.1 | 5 | 475 | |||
GenBank mRNA | AF087861.1 | - | 4 | 256 | ||
AK302147.1 | - | 6 | 891 |
Gene Symbol: RAB4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016154.4 | NP_057238.3 | 3 | 270 | ||
GenBank mRNA | AF165522.1 | - | 3 | 236 | ||
AF217985.1 | - | 2 | 1074 | |||
AF498935.1 | - | 3 | 113 | |||
BC033081.1 | - | 3 | 181 | 5456088 | ||
BC046927.1 | - | 3 | 223 | 5805921 | ||
BG706803.1 | - | 3 | 278 |