Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317724.1 | NP_001304653.1 | 4, 5 | 443 | ||
NM_016167.4 | NP_057251.2 | 4, 5 | 443 | |||
GenBank mRNA | AF172066.3 | - | 4, 5 | 443 | ||
AK075301.1 | - | 4, 5 | 426 | |||
AL050299.1 | - | 3, 4 | 143 | |||
BC023517.2 | - | 4, 5 | 418 | 4298095 | ||
BC062683.1 | - | 4, 5 | 439 | 6499123 | ||
BU071935.1 | - | 4, 5 | 196 |