Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016172.2 | NP_057256.2 | 2, 3 | 478 | ||
GenBank mRNA | AF038200.1 | - | 1, 2 | 43 | ||
AF068195.1 | - | 1, 2 | 105 | |||
AF176796.1 | - | 2, 3 | 451 | |||
BC004967.1 | - | 2, 3 | 478 | 3535682 | ||
BC011822.1 | - | 2, 3 | 304 | 3688526 |