Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016050.4 | NP_057134.1 | 2 | 276 | ||
NM_170738.3 | NP_733934.1 | 2 | 198 | |||
NM_170739.3 | NP_733935.1 | 2 | 276 | |||
NR_120588.1 | 2 | 276 | ||||
NR_120589.1 | 2 | 126 | ||||
XM_017018135.1 | XP_016873624.1 | 2 | 210 | |||
GenBank mRNA | AB049638.1 | - | 2 | 248 | ||
AF151871.1 | - | 2 | 260 | |||
AK127603.1 | - | 2 | 885 | |||
AK223112.1 | - | 2 | 252 | |||
AK290084.1 | - | 2 | 254 | |||
BC005002.1 | - | 2 | 255 | 3542758 | ||
BC108277.1 | - | 2 | 171 | 4416083 | ||
BI193255.1 | - | 2 | 190 | |||
BM554886.1 | - | 2 | 238 | |||
BM755421.1 | - | 2 | 267 | |||
BU597133.1 | - | 2 | 279 |