Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLRX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243399.1 | NP_001230328.1 | 4 | 904 | ||
NM_001319291.1 | NP_001306220.1 | 4 | 485 | |||
NM_016066.4 | NP_057150.2 | 4 | 1062 | |||
NM_197962.2 | NP_932066.1 | 4 | 679 | |||
XM_017001397.1 | XP_016856886.1 | 4 | 612 | |||
GenBank mRNA | AF132495.2 | - | 4 | 597 | ||
AF151891.1 | - | 4 | 569 | |||
AF290514.1 | - | 4 | 1062 | |||
AY038988.1 | - | 4 | 601 | |||
BC028113.1 | - | 4 | 585 | 5395272 | ||
BG182176.1 | - | 3 | 471 |