Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PALLD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166108.1 | NP_001159580.1 | 22 | 4263 | ||
NM_001166109.1 | NP_001159581.1 | 19 | 3331 | |||
NM_001166110.1 | NP_001159582.1 | 12 | 2830 | |||
NM_016081.3 | NP_057165.3 | 21 | 4212 | |||
XM_005262861.4 | XP_005262918.1 | 22 | 4916 | |||
XM_005262866.2 | XP_005262923.1 | 21 | 4466 | |||
XM_011531768.2 | XP_011530070.1 | 23 | 5170 | |||
XM_011531769.2 | XP_011530071.1 | 22 | 5119 | |||
XM_011531770.2 | XP_011530072.1 | 24 | 5216 | |||
XM_011531771.2 | XP_011530073.1 | 21 | 4897 | |||
XM_011531772.2 | XP_011530074.1 | 20 | 4798 | |||
XM_011531773.1 | XP_011530075.1 | 22 | 4498 | |||
XM_011531774.1 | XP_011530076.1 | 21 | 4447 | |||
XM_011531775.1 | XP_011530077.1 | 21 | 3851 | |||
XM_017007910.1 | XP_016863399.1 | 23 | 5165 | |||
GenBank mRNA | AB023209.1 | - | 12 | 2793 | ||
AF077041.1 | - | 8 | 2279 | |||
AF151909.1 | - | 8 | 2185 | |||
AF464873.1 | - | 21 | 4211 | |||
AL050093.1 | - | 3 | 914 | |||
BX537391.1 | - | 5 | 1113 |