Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286045.1 | NP_001272974.1 | 5 | 1406 | ||
NM_001286046.1 | NP_001272975.1 | 5 | 1402 | |||
NM_001286047.1 | NP_001272976.1 | 4 | 1243 | |||
NM_001286048.1 | NP_001272977.1 | 3 | 1155 | |||
NM_016090.3 | NP_057174.1 | 5 | 1403 | |||
GenBank mRNA | AB209753.1 | - | 4 | 2949 | ||
AF156098.1 | - | 5 | 1281 | |||
AK002015.1 | - | 5 | 1319 | |||
AK225615.1 | - | 5 | 1272 | |||
AK225667.1 | - | 5 | 1297 | |||
BC034381.1 | - | 5 | 1270 | 5180771 | ||
BC070262.1 | - | 5 | 1264 | 4619267 |