Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF706 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042510.1 | NP_001035975.1 | 4, 5 | 655 | ||
NM_001267708.1 | NP_001254637.1 | 5, 6 | 699 | |||
NM_001267709.1 | NP_001254638.1 | 3, 4 | 453 | |||
NM_016096.3 | NP_057180.1 | 3, 4 | 540 | |||
GenBank mRNA | AF125099.1 | - | 3, 4 | 376 | ||
AF275802.1 | - | 3, 4 | 421 | |||
AF275809.1 | - | 3, 4 | 421 | |||
AK027802.1 | - | 3, 4 | 1764 | |||
AK290477.1 | - | 3, 4 | 424 | |||
BC015925.1 | - | 1 | 297 | 3909510 | ||
BC068524.1 | - | 3, 4 | 424 | 5263977 | ||
BG704587.1 | - | 3, 4 | 458 | |||
BI755311.1 | - | 4, 5 | 563 | |||
DB231109.1 | - | 3, 4 | 454 |