Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016107.3 | NP_057191.2 | 20 | 4153 | ||
GenBank mRNA | AF100742.1 | - | 10 | 2149 | ||
AK025481.1 | - | 8 | 1846 | |||
AK055938.1 | - | 3 | 1902 | |||
AK123306.1 | - | 4 | 2550 | |||
AK307808.1 | - | 6 | 1492 | |||
AL137258.1 | - | 3 | 1168 | |||
BC051893.1 | - | 13 | 2743 | |||
BC062986.1 | - | 13 | 2668 |