Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COPS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258006.1 | NP_001244935.1 | 7 | 970 | ||
NM_016129.2 | NP_057213.2 | 7 | 970 | |||
XM_011532019.1 | XP_011530321.1 | 7 | 966 | |||
GenBank mRNA | AF100757.1 | - | 7 | 834 | ||
AK001148.1 | - | 7 | 966 | |||
AK001210.1 | - | 7 | 797 | |||
AK024005.1 | - | 7 | 855 | |||
AK094238.1 | - | 7 | 1608 | |||
AK223178.1 | - | 7 | 840 | |||
BC004302.1 | - | 7 | 833 | 3623514 | ||
BC009292.2 | - | 7 | 833 | 4111685 | ||
BC093007.1 | - | 7 | 855 | 30340740 | ||
KU178633.1 | - | 7 | 831 | |||
KU178634.1 | - | 7 | 831 |