Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUFU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016169.3 | NP_057253.2 | 12 | 4518 | ||
XM_011539858.2 | XP_011538160.1 | 13 | 4647 | |||
XM_011539860.2 | XP_011538162.1 | 13 | 4644 | |||
XM_011539861.2 | XP_011538163.1 | 12 | 4521 | |||
XM_011539863.2 | XP_011538165.1 | 13 | 4382 | |||
XM_017016323.1 | XP_016871812.1 | 13 | 4877 | |||
GenBank mRNA | AF159447.1 | - | 12 | 4473 | ||
AL137465.1 | - | 2 | 3006 |