Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016172.2 | NP_057256.2 | 10 | 1627 | ||
GenBank mRNA | AF038200.1 | - | 9 | 1192 | ||
AF068195.1 | - | 9 | 1254 | |||
AF176796.1 | - | 10 | 1597 | |||
BC004967.1 | - | 10 | 1627 | 3535682 | ||
BC011822.1 | - | 10 | 1453 | 3688526 | ||
BX647463.1 | - | 3 | 5680 |