Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dctn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198866.1 | NP_001185795.1 | 21 | 2404 | ||
NM_001198867.1 | NP_001185796.1 | 18 | 2344 | |||
NM_007835.2 | NP_031861.2 | 21 | 2650 | |||
GenBank mRNA | AK147769.1 | - | 14 | 1824 | ||
AK157867.1 | - | 21 | 2651 | |||
AK171376.1 | - | 18 | 2331 | |||
AK172261.1 | - | 18 | 2318 | |||
AK202724.1 | - | 2 | 71 | |||
BC066061.1 | - | 21 | 2377 | 6849060 | ||
U60312.1 | - | 21 | 2385 |