Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dctn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198866.1 | NP_001185795.1 | 19, 20 | 2290 | ||
NM_001198867.1 | NP_001185796.1 | 16, 17 | 2230 | |||
NM_007835.2 | NP_031861.2 | 19, 20 | 2536 | |||
GenBank mRNA | AK157867.1 | - | 19, 20 | 2537 | ||
AK171376.1 | - | 16, 17 | 2217 | |||
AK172261.1 | - | 16, 17 | 2204 | |||
BC066061.1 | - | 19, 20 | 2263 | 6849060 | ||
U60312.1 | - | 19, 20 | 2271 |